Cole-Parmer Intelligent Touch-Screen Rotary Viscometer; 10 to 2,000,000 mPa·s
Representative image only.
Cole-Parmer Intelligent Touch-Screen Rotary Viscometer; 10 to 2,000,000 mPa·s
Cole-Parmer | Item: 8305597
Enable multiple sampling in one rotation
₹594,000.00INR / Each
for an additional
₹43,600.00INR / Each
for an additional
₹65,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹594,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Intelligent Touch-Screen Rotary Viscometer; 10 to 2,000,000 mPa·s
Cole-Parmer Intelligent Touch-Screen Rotary Viscometer; 10 to 2,000,000 mPa·s
8305597
₹594,000.00INR / Each
+
InnoCal NIST-Traceable Calibration; Viscometer
1710701
₹43,600.00INR / Each
+ =
Total Price 915600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Spindles, lifting rod with pedestal, power adapter, protection framework, wrench, and RTD temperature probe.
Key Features
  • Improved touch-screen interface
  • Anti-static shell and PC material lifting pole
  • Durable new design with smaller shaft
  • ARM chip processor for higher data processing speed
  • RTD temperature probe monitors sample temperature in real time
More About this Item
Intelligent touch-screen rotary viscometers are ideal for measurement of viscous resistance and absolute viscosity of liquid. This is widely applied in the field of petri chemistry, medicine, food, light industry, textile industry, scientific research, and more. The 5" touch-screen is easy to use and shows multiple parameters at once such as viscosity, speed, torque percentage, max measurement range under the current rotor, and speed. They can convert freely between various viscosity units automatically. The professional model (83055-98) features a larger 7" screen. RS-232 connection to a computer allows the user to constantly control rotation speed to maximize their process. Results can then be easily analyzed and printed. The professional model (83055-98) may also be used via USB interface.
Note: To read 1Mpas, a 0 spindle is required. The viscometer must be ordered with the 0 spindle to provide the required linear curve programming. Please contact us for more details.
1 Year Warranty
Specifications
  • Viscosity10 to 2,000,000 mPa·s
  • Repeatability±0.5% (Newtonian Fluids)
  • Display5" touch-screen
  • OutputRS-232
  • Rpm0.3, 0.6, 1.5, 3, 6, 12, 30, 60
  • Power (VAC)100 to 240
  • DescriptionIntelligent Touch-Screen Rotary Viscometer; 10 to 2,000,000 mPa·s