InnoCal A2LA ISO17025 Accredited Calibration; Viscometer
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration; Viscometer
InnoCal | Item: 1750119
Ensure your instrument remains precise
₹65,700.00INR / Each
₹65,700.00INR / Each
Buy It Together
Current Item
InnoCal A2LA ISO17025 Accredited Calibration; Viscometer
InnoCal A2LA ISO17025 Accredited Calibration; Viscometer
1750119
₹65,700.00INR / Each
+ =
Total Price 138300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • A2LA Accredited Calibration. View our scope of accreditation
  • Calibration service for a viscometer
More About this Item
Rotational viscometers are tested at the midpoint value of the spindles. Accuracy is confirmed using ISO Guide 34 Accredited Standard Reference Materials.
Specifications
  • Calibration TypeA2LA ISO17025 Accredited Liquid Testing
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points1 Per Spindle
  • Calibration Test PointsApproximately Mid-Point of Range
  • DescriptionA2LA ISO17025 Accredited Calibration; Viscometer