Cole-Parmer In-Line pH Probe, DJ/PVDF/1kOhm RTD/10'; Spade/Tinned
Representative image only.
Cole-Parmer In-Line pH Probe, DJ/PVDF/1kOhm RTD/10'; Spade/Tinned
Cole-Parmer | Item: 0599348
Integral, one-piece electrode and housing for installation convenience
₹38,100.00INR / Each
₹38,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer In-Line pH Probe, DJ/PVDF/1kOhm RTD/10'; Spade/Tinned
Cole-Parmer In-Line pH Probe, DJ/PVDF/1kOhm RTD/10'; Spade/Tinned
0599348
₹38,100.00INR / Each
+ + =
Total Price 255000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from single-junction or double-junction designs
  • Options of BNC or stripped and tinned ends
  • 1 year warranty
More About this Item
In-line electrodes operate from 32 to 212°F (0 to 100°C) and have a chemical-resistant PVDF housing with a 1/2'' NPT(M) thread. Housing measures 4'' x 15/16'' diameter. Electrode extends 1'' beyond housing. Maximum pressure 100 psi. Measure pH from 0 to 13.
Note: 100 Ω RTD ATC elements are platinum and conform to DIN curve 43760. ATC connector is miniature audio plug.
Connector for the 1 kΩ RTD and the 3 kΩ Balco ATC elements is a spade lug.
Connector for 100 kΩ
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialPDVF
  • Min pH (pH)0
  • Max pH (pH)13
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • ATC ConnectorSpade lugs
  • ATC Element1 kΩ Pt RTD
  • ConnectorTinned ends
  • DescriptionIn-Line pH Probe, DJ/PVDF/1kOhm RTD/10'; Spade/Tinned