Cole-Parmer HDPE In-Line Filter, Polypropylene, Natural, 90-130 Micron, 1/4" ID; 10/Pk
Representative image only.
Cole-Parmer HDPE In-Line Filter, Polypropylene, Natural, 90-130 Micron, 1/4" ID; 10/Pk
Cole-Parmer | Item: 5014134
Effectively removes visible particles, impurities, and contaminants
₹1,900.00INR / Pkg of 10
₹1,900.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer HDPE In-Line Filter, Polypropylene, Natural,  90-130 Micron, 1/4" ID; 10/Pk
Cole-Parmer HDPE In-Line Filter, Polypropylene, Natural, 90-130 Micron, 1/4" ID; 10/Pk
5014134
₹1,900.00INR / Pkg of 10
+ + + + + + + =
Total Price 305100.00 ADD 8 ITEMS TO CART
Product Description
Key Features
  • Suitable for a broad range of industrial and commercial applications
  • Built with a robust polypropylene housing for long-lasting performance
  • Equipped with hose barb fittings to fit various tubing sizes
More About this Item
These HDPE in-line filters are engineered to effectively remove visible particles, impurities, and contaminants from fluid systems. Suitable for a wide range of industries, these filters provide reliable performance across diverse applications.
Each filter features a durable polypropylene housing with hose barb fittings on both ends, ensuring compatibility with various tubing sizes. Available in 45–90 micron and 90–130 micron options, they offer versatile filtration to meet your specific needs.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Membrane MaterialHDPE
  • Housing MaterialPolypropylene (PP)
  • ColorNatural
  • Filter Size90-130 Micron
  • Connections1/4" ID Hose Barb
  • DescriptionHDPE Inline Filter, Polypropylene, Natural, 90-130 Micron, 1/4" ID; 10/Pk