KD Scientific KDS 100 Legacy 100 Single-Syringe Infusion Pump
Easy-to-use syringe pump delivers high precision and excellent durability
  • Accepts a single syringe with 10 µL to 60 mL capacity
  • Quickly set parameters using menu-driven setup
  • Precise flow control with less than ±1% accuracy
  • Dispense up to 423 mL/hr (with a 60 mL syringe)
The single-syringe infusion pump combines precision with simplicity to deliver accurate amount of fluid every time. Pump accommodates a syringe ranging from 10 µL to 60 mL. A button release of drive nut offers easy syringe installation.
Simple menu-driven setup makes inputting parameters quick and easy: select syringe size from the built-in syringe size table, set dispense volume, then set dispense flow rate. Volume and flow rate units (µl/hr or ml/hr) are selected automatically based on syringe size. View dispensing volume on the easy-to-read backlit LCD. Meter automatically shuts off at completion. Last setting stored in permanent memory for your convenience.
258,000.00 - ₹284,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Syringe (mL/min) Min Flow Rate Per Syringe (picoL/min) Power (VAC)
Availability Pricing
KD Scientific KDS 100 Legacy 100 Single-Syringe Infusion Pump; 110V 5100510 Mfr # 780100
COMPARE
7.05 1666.7 110
₹258,000.00INR / Each
KD Scientific KDS 100 Legacy 100 Single-Syringe Infusion Pump; 220V 5100511 Mfr # 789100
COMPARE
7.05 1666.7 220
₹284,000.00INR / Each