Cole-Parmer HandE-Vac Safety Stand for Hand Operator and Adapters
Representative image only.
Cole-Parmer HandE-Vac Safety Stand for Hand Operator and Adapters
Cole-Parmer | Item: 0439605
Optimize performance of your E-Vac aspiration system
₹19,300.00INR / Each
₹19,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer HandE-Vac Safety Stand for Hand Operator and Adapters
Cole-Parmer HandE-Vac Safety Stand for Hand Operator and Adapters
0439605
₹19,300.00INR / Each
+
Cole-Parmer MiniVac Waste Aspiration System, 1 L
0439619
₹11,800.00INR / Each
+
Cole-Parmer Heavy-Duty Plastic Bottle, PP, 2000mL; 2/PK
0625720
₹16,200.00INR / Pkg of 2
+ + + + =
Total Price 228030.00 ADD 7 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Accessories for E-Vac system designed to collect critical or dangerous liquids
More About this Item
Make the most of your aspiration system with extra bottles, replacement parts, and other accessories. E-Vac aspiration systems available as 04397-10 and 13050-31, sold separately.
Specifications
  • DescriptionSafety Stand for Hand Operator and Adapters