Cole-Parmer Glass Capillary Tubes, Closed Both Ends; 100/PK
Representative image only.
Cole-Parmer Glass Capillary Tubes, Closed Both Ends; 100/PK
Cole-Parmer | Item: 0301365
Accessories for your Cole-Parmer melting point apparatus
Call For Price
Call For Price
Product Description
Key Features
  • Constructed of soda glass and easily sealed in a Bunsen flame
  • Supplied in a durable tube holder for added protection
  • Tubes are closed on both ends
  • Previous part number SMP2/1
More About this Item
Capillary tubes measure 100 mm L, 1.9 mm OD, 1.3 mm ID, and 0.3 mm wall thickness.
Specifications
  • MaterialGlass
  • Compatible With03011-40, 03011-41, 03011-45, 03011-47, 03011-49, 03013-02, 03013-03, 03013-60, 03013-61
  • Length (mm)100
  • Diameter (mm)1.9
  • DescriptionGlass Capillary Tubes, Closed at Both Ends; 100/PK