Cole-Parmer FSB-200-D Compact Fluidized Sand Bath, 50 to 600 °C, 4 x 1000 W; 230 VAC

Representative image only.

Cole-Parmer FSB-200-D Compact Fluidized Sand Bath, 50 to 600 °C, 4 x 1000 W; 230 VAC

Cole-Parmer | Item: 0118950

Fast heat-ups and accurate temperature control to 600°C

Call For Price

Call For Price

Buy It Together

Current Item

Cole-Parmer FSB-200-D Compact Fluidized Sand Bath, 50 to 600 °C, 4 x 1000 W; 230 VAC
Cole-Parmer FSB-200-D Compact Fluidized Sand Bath, 50 to 600 °C, 4 x 1000 W; 230 VAC
0118950

Call For Price
+ +
Cole-Parmer Lid for Industrial Fluidized Sand Baths
0118452

₹130,000.00INR / Each
+
Cole-Parmer Brown Aluminum Oxide, #120 Grit, 2x50lbs
0118461

₹69,700.00INR / Pkg
=
Total Price 615700.00 ADD 4 ITEMS TO CART
Product Description

Includes

Overspill flange and power cord with plug.

Key Features

  • Dry, inert, nontoxic aluminum oxide ensures hazard-free operation
  • Safer alternative to high temperature oil and salt baths
  • Requires controller (01184-50) in order to be calibrated
  • Heats up from 20 °C to maximum in 105 minutes
  • Requires 100 lb (45.4 kg) of brown sand 01184-61
  • 3 year warranty
  • Previous part number F946D

More About this Item

Use these fluidized sand baths for your heat treating processes, temperature sensor calibrations, and electrical component testing. Baths use aluminum oxide fluidized by low-pressure air as a dry bath medium with excellent heat transfer properties. Inert, nonflammable aluminum oxide is safer to use than oil and will not corrode your sample vessels or produce toxic fumes. Vessels of all shapes can be easily immersed and safely removed from the bath, without the hazard and mess of hot oils.

Make temperature adjustments via a control knob and reference dial. Baths require a dry air flow of approximately 2 cu ft/min (57 L/min) at about 3 psi pressure; order an air pump separately. Order temperature controller (01184-50 or 01184-55) to accurately control the temperature of your sand bath. Baths feature a thermowell to hold a thermocouple thermometer.

Note: not recommended for shape setting in laboratory research device manufacturing or for heating of reactors.

Working dimensions: 13-3/4" deep x 9" dia (35 x 22.8 cm)
Overall dimensions (excluding tap): 27-3/8"H x 15-3/16" dia (69.5 x 38.5 cm)

Please Note: Effective June 2022, the Techne Fluidized Sand Bath Model number F946D has now been rebranded Cole-Parmer under Model number FSB-200-D. All the same great features and specifications, just with a new and improved look.

3 Year Warranty
Specifications
  • Bath TypeSand Baths
  • Max Temperature (° C)600
  • Min Temperature (° C)50
  • Temperature Stability (° C)±1.0 at 50°C; 3°C at 600°C
  • Height (in)27 3/8
  • Height (cm)69.5
  • Working Area Depth (in)13 3/4
  • Heater Wattage (watts)1000
  • Wetted MaterialsStainless steel
  • Power (VAC)230
  • Power (Hz)50/60
  • DescriptionCompact Fluidized Sand Bath, 50 to 600 °C, 4 x 1000 W; 230 VAC
  • Warranty3 years
Reviews
Q&A