Cole-Parmer Lid for Industrial Fluidized Sand Baths
Representative image only.
Cole-Parmer Lid for Industrial Fluidized Sand Baths
Cole-Parmer | Item: 0118452
₹130,000.00INR / Each
₹130,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Lid for Industrial Fluidized Sand Baths
Cole-Parmer Lid for Industrial Fluidized Sand Baths
0118452
₹130,000.00INR / Each
+ + + =
Total Price 809000.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Insulated lid help maintain bath temperature
  • Previous part number F6156
More About this Item
Parts and accessories to meet your application needs and keep your sand bath operating dependably.
Use a lid to contain aluminum oxide and retain heat. Unique design allows air to flow into the top of the bath and then through the exhaust duct when connected to an extraction system.
Specifications
  • Compatible WithIndustrial fluidized sand baths 01184-10 and 01184-20
  • DescriptionLid for Industrial Fluidized Sand Baths