Cole-Parmer Flow-Through Tubing Weight, PP, 5/16 to 7/16" ID Tubing
Representative image only.
Cole-Parmer Flow-Through Tubing Weight, PP, 5/16 to 7/16" ID Tubing
Cole-Parmer | Item: 4852554
Prevent tubing from coiling or floating during fluid transfer applications
₹13,400.00INR / Each
₹13,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Flow-Through Tubing Weight, PP, 5/16 to 7/16" ID Tubing
Cole-Parmer Flow-Through Tubing Weight, PP, 5/16 to 7/16" ID Tubing
4852554
₹13,400.00INR / Each
+
Tygon A-60-F Tubing, 1/8" ID x 1/4" OD; 50 Ft
5010600
Call For Price
=
Total Price 13400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Tapered hose barb fitting easily inserts into the inner diameter (ID) of flexible tubing
  • Keep tubing submerged in liquid, preventing flotation, and ensuring uninterrupted fluid flow
  • Autoclavable—allowing for easy sterilization and reuse
  • Suitable for tubing with 5/16" to 7/16" inner diameter
  • Constructed from chemically resistant PP
  • Resin is FDA-compliant and meets USP Class VI requirements
More About this Item
Cole-Parmer® flow-through tubing weights provide the perfect solution for keeping tubing submerged, preventing floatation, and ensuring uninterrupted flow throughout the fluid transfer process. Crafted from high-quality materials, these tubing weights ensure compatibility with a wide range of fluids and preventing contamination. Weighted ends are open in the center and feature a tapered hose barb fitting for easy attachment to flexible tubing , ensuring a snug and secure fit. Designed for maximum convenience and versatility, our tubing weights are autoclavable at 121°C for 20 minutes at 15 psi.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Minimum ID (InchShort)0.3125
  • Width (InchShort)9/16
  • Maximum ID (InchShort)0.4375
  • Minimum ID (mm)7.9375
  • Height (InchShort)1 19/32
  • Maximum ID (mm)11.1125
  • MaterialPP
  • DescriptionFlow Through Tubing Weight, PP, 5/16" to 7/16" Tubing