Cole-Parmer® Flow-Through Tubing Weight
Prevent tubing from coiling or floating during fluid transfer applications
  • Tapered hose barb fitting easily inserts into the inner diameter (ID) of flexible tubing
  • Keep tubing submerged in liquid, preventing flotation, and ensuring uninterrupted fluid flow
  • Autoclavable—allowing for easy sterilization and reuse
Cole-Parmer® flow-through tubing weights provide the perfect solution for keeping tubing submerged, preventing floatation, and ensuring uninterrupted flow throughout the fluid transfer process. Crafted from high-quality materials, these tubing weights ensure compatibility with a wide range of fluids and preventing contamination. Weighted ends are open in the center and feature a tapered hose barb fitting for easy attachment to flexible tubing , ensuring a snug and secure fit. Designed for maximum convenience and versatility, our tubing weights are autoclavable at 121°C for 20 minutes at 15 psi.
12,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Maximum ID (InchShort) Minimum ID (InchShort) Description
Availability Pricing
COMPARE
0.25 0.1875 Flow-Through Tubing Weight, PTFE, 3/16" to 1/4" Tubing
₹12,200.00INR / Each
COMPARE
0.4375 0.3125 Flow-Through Tubing Weight, PTFE, 5/16" to 7/16" Tubing
₹12,200.00INR / Each