Cole-Parmer FHD-400 Carbon Filter for Inorganic Vapors
Representative image only.
Cole-Parmer FHD-400 Carbon Filter for Inorganic Vapors
Cole-Parmer | Item: 7890236
A filter for every application
₹133,000.00INR / Each
₹133,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer FHD-400 Carbon Filter for Inorganic Vapors
Cole-Parmer FHD-400 Carbon Filter for Inorganic Vapors
7890236
₹133,000.00INR / Each
+
Mark-10 M2-2 Force Gauge, 2 lbF / 1 kgF / 10 N
2530272
₹140,000.00INR / Each
+ + + +
Cole-Parmer Airflow Meter Vaneometer
7890012
₹22,900.00INR / Each
+ + + =
Total Price 3971900.00 ADD 9 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Remove pollutants safety
  • Main filter for inorganic vapors including acids
  • Units require qty 2 each
  • 1 year warranty
More About this Item
Choose from a wide range of filters to remove a wide variety of pollutants. Filters are compatible with the Cole-Parmer range of ductless fume hoods
1 Year Warranty
Specifications
  • Filter TypeCarbon for inorganic vapors
  • DescriptionFHD-400 Ductless Fume Hood Carbon Filter for Inorganic Vapors including acids, units required qty 2 each.