Cole-Parmer Electronic Ice Bucket Spare Bucket
Representative image only.
Cole-Parmer Electronic Ice Bucket Spare Bucket
Cole-Parmer | Item: 9996793
Keep your samples cold, clean, and dry
₹18,900.00INR / Each
₹18,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer Electronic Ice Bucket Spare Bucket
Cole-Parmer Electronic Ice Bucket Spare Bucket
9996793
₹18,900.00INR / Each
+ =
Total Price 240900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
These Techne N°ICE ceramic-coated beads maintain the temperature and support the sample vessels for as long as necessary. The beads are chemically resistant and if required can be autoclaved at 134°C. As a time saving option, the bucket of beads can be incubated in the fridge overnight and simply inserted into the unit as and when required. Compatible with the 24120-00 Electronic Ice Bucket, sold separately.
1 Year Warranty
Specifications
  • DescriptionElectronic Ice Bucket Spare Bucket