Cole-Parmer DO6050-T DO Probe Maintenance Kit, for 35201-80
Representative image only.
Cole-Parmer DO6050-T DO Probe Maintenance Kit, for 35201-80
Cole-Parmer | Item: 3520190
Get the most out of water analysis equipment
₹18,000.00INR / Each
₹18,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer DO6050-T DO Probe Maintenance Kit, for 35201-80
Cole-Parmer DO6050-T DO Probe Maintenance Kit, for 35201-80
3520190
₹18,000.00INR / Each
+
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Transmitter
0580214
₹46,300.00INR / Each
+ + + =
Total Price 228200.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Replace or upgrade electrodes, probes, cells, and other accessories
  • 1 year warranty
More About this Item
Water analysis accessories for Cole-Parmer® meters. Many of these items are compatible with other brands of meter.
Includes five membranes, five O-rings, five spacers, and 250-mL of electrolyte solution.
1 Year Warranty
Specifications
  • Compatible With35201-80
  • DescriptionDO Probe Maintenance Kit, for 35201-80