Cole-Parmer DLF-400 Horizontal Laminar Flow Cabinet, 72" Width; 230V, 50Hz

Representative image only.

Cole-Parmer DLF-400 Horizontal Laminar Flow Cabinet, 72" Width; 230V, 50Hz

Cole-Parmer | Item: 7890253

Provides reliable protection for samples and work processes from particulates

₹1,085,000.00INR / Each

₹1,085,000.00INR / Each

Buy It Together

Current Item

Cole-Parmer DLF-400 Horizontal Laminar Flow Cabinet, 72" Width; 230V, 50Hz
Cole-Parmer DLF-400 Horizontal Laminar Flow Cabinet, 72" Width; 230V, 50Hz
7890253

₹1,085,000.00INR / Each
+ + =
Total Price 1454000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • ULPA filtration technology creates a clean work environment over a wide range of applications
  • Meets and exceeds ISO Class 4 compliance for clean air
  • Horizontal flow of filtered air does not directly impinge on the work surface, reducing turbulence
  • Constructed with powder-coated interior and stainless steel work surface
  • High-capacity air handling system delivers flow velocity of 0.4 m/s to 0.5 m/s (80 to 100 fpm)
  • Operates on 230 VAC, 50 Hz power
  • 3 year warranty

More About this Item

These Cole-Parmer® DLF-400 horizontal laminar flow cabinets are designed to protect equipment and other contents from particulates. Airflow from horizontal flow cabinets flows from back to front and does not impinge on the work surface but is smoothly drawn across it. The ULPA filter is mounted behind the work surface, providing a stream of clean, uniform air within the work zone.

Laminar flow cabinets use ULPA filters which are 99.999% efficient at particle sizes between 0.1 to 0.3 μm along with a disposable polyester fiber pre-filter with 85% arrestance. All models feature a stainless steel work surface with spill retention, compact LED cabinet lamp, side “viewing” windows, and control panel. Side windows allow ambient light into the chamber and provide users with three points of visibility for an unobstructed view of contents. The control panel features on/off switches for the interior lamp, blower, and electrical outlet, and a pressure gauge to measure filter performance.

Cabinets are ideally suited for use with non-hazardous contaminants when flexible access to equipment in the work zone is desired. These are suitable for applications that generate no biohazardous materials and do not require operator protection. Designed for desktop use or installation on the optional mobile base stand. Base stand and UV lamp are sold separately as optional accessories.

Please note: UV lamps must be purchased at the same time as the compatible cabinet since control panel with a UV key is factory installed in the cabinet.

Conformite EuropeenneCanadian Standards Association3 Year Warranty
Specifications
  • Hood Width (in)72
  • Blower Typeebm-papst external rotor motor
  • LightsLED
  • Exterior Width (in)73.25
  • Exterior Height (in)42.75
  • Exterior Depth (in)29.5
  • Interior Width (in)72
  • Interior Height (in)23.75
  • Interior Depth (in)23.75
  • Power (VAC)230
  • Power (Hz)50
  • Shipping Weight (lbs)595
  • Shipping Weight (kg)270
  • Filter TypeULPA main filter and polyester fiber pre-filter
  • Face Velocity Accuracy0.4 to 0.5 m/s (80 to 100 fpm)
  • DescriptionHorizontal Laminar Flow Cabinet, 72" Width; 230V, 50Hz
  • Warranty3 Years
Reviews
Q&A