Cole-Parmer DC-400 Desiccator Cabinet, Aluminum Frame, Two Shelves; 70 L

Representative image only.

Cole-Parmer DC-400 Desiccator Cabinet, Aluminum Frame, Two Shelves; 70 L

Cole-Parmer | Item: 7890281

Safely store humidity sensitive products

₹159,000.00INR / Each

₹159,000.00INR / Each

Buy It Together

Current Item

Cole-Parmer DC-400 Desiccator Cabinet, Aluminum Frame, Two Shelves; 70 L
Cole-Parmer DC-400 Desiccator Cabinet, Aluminum Frame, Two Shelves; 70 L
7890281

₹159,000.00INR / Each
+
Traceable Monitoring Hygrometer
2025030

₹6,700.00INR / Each
+
Cole-Parmer Indicating Desiccant, 8 Mesh; 2.5kg
8840159

Call For Price
+ +
Drierite 12005 Nonindicating Desiccant,  6 Mesh; 5 lb Jar, 4/CS
0893413

₹54,500.00INR / Case of 4
+
Boekel Shelf for Static-Free Desiccators
2400050

₹106,000.00INR / Each
=
Total Price 360000.00 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Two SS shelves and a key.

Key Features

  • Easily view your samples through acrylic viewing panels
  • Durable, rugged construction— aluminum frame and stainless steel (SS) shelves
  • SS shelves with holes ensure uniform humidity throughout chamber
  • 2.47 cu ft (70 L) capacity
  • 3 year warranty

More About this Item

Desiccators provide dust-free storage for humidity sensitive products such as optical equipment, electronic parts, PCB boards, and reagents. Place a desiccant material in the cabinets to keep the environment dry. Once the desiccant is saturated, simply replace or regenerate the desiccant for reuse. A lock and key let you securely lock the cabinet to prevent unauthorized access.

3 Year Warranty
Specifications
  • Chamber Volume (cu ft)2.47
  • Chamber Volume (Liters)70
  • Interior Width (in)12.2
  • Interior Height (in)15.9
  • Interior Depth (in)18.8
  • Interior Width (cm)31
  • Interior Height (cm)40.5
  • Interior Depth (cm)47.8
  • Exterior Width (in)15
  • Exterior Height (InchShort)22.9
  • Exterior Depth (in)19.8
  • Exterior Width (cm)38
  • Exterior Height (cm)58.1
  • Exterior Depth (cm)50.2
  • MaterialAluminum frame, acrylic panels, and SS shelves
  • ColorClear
  • DescriptionDesiccator Cabinet, Aluminum Frame, Two Shelves; 70 L
  • Warranty2 Years
Reviews
Q&A