Cole-Parmer DC-400 Desiccator Cabinet, Aluminum Frame, One Shelf; 50 L

Representative image only.

Cole-Parmer DC-400 Desiccator Cabinet, Aluminum Frame, One Shelf; 50 L

Cole-Parmer | Item: 7890280

Safely store humidity sensitive products

₹196,000.00INR / Each

₹196,000.00INR / Each

Buy It Together

Current Item

Cole-Parmer DC-400 Desiccator Cabinet, Aluminum Frame, One Shelf; 50 L
Cole-Parmer DC-400 Desiccator Cabinet, Aluminum Frame, One Shelf; 50 L
7890280

₹196,000.00INR / Each
+ +
Drierite 12005 Nonindicating Desiccant,  6 Mesh; 5 lb Jar, 4/CS
0893413

₹54,500.00INR / Case of 4
+
Traceable Monitoring Hygrometer
2025030

₹6,700.00INR / Each
+
Boekel Shelf for Static-Free Desiccators
2400050

₹106,000.00INR / Each
+
Cole-Parmer Indicating Desiccant, 8 Mesh; 500g
8840160

Call For Price
=
Total Price 397000.00 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Includes

One SS shelf and a key.

Key Features

  • Easily view your samples through acrylic viewing panels
  • Durable, rugged construction— aluminum frame and stainless steel (SS) shelves
  • SS shelves with holes ensure uniform humidity throughout chamber
  • 1.76 cu ft (50 L) capacity
  • 3 year warranty

More About this Item

Desiccators provide dust-free storage for humidity sensitive products such as optical equipment, electronic parts, PCB boards, and reagents. Place a desiccant material in the cabinets to keep the environment dry. Once the desiccant is saturated, simply replace or regenerate the desiccant for reuse. A lock and key let you securely lock the cabinet to prevent unauthorized access.

3 Year Warranty
Specifications
  • Chamber Volume (cu ft)1.76
  • Chamber Volume (Liters)50
  • Interior Width (in)13
  • Interior Height (in)13.2
  • Interior Depth (in)13
  • Interior Width (cm)33
  • Interior Height (cm)33.5
  • Interior Depth (cm)33
  • Exterior Width (in)15.7
  • Exterior Height (InchShort)17
  • Exterior Depth (in)17
  • Exterior Width (cm)40
  • Exterior Height (cm)43.3
  • Exterior Depth (cm)43.3
  • MaterialAluminum frame, acrylic panels, and SS shelf
  • ColorClear
  • DescriptionDesiccator Cabinet, Aluminum Frame, One Shelf; 50 L
  • Warranty2 Years
Reviews
Q&A