Cole-Parmer Cleanroom Foam Swab, Open-Hole Head, Notched, Light Green PP Handle; 500/PK
Representative image only.
Cole-Parmer Cleanroom Foam Swab, Open-Hole Head, Notched, Light Green PP Handle; 500/PK
Cole-Parmer | Item: 3367605
Ideal for precision cleaning
₹19,700.00INR / Pkg of 500
₹19,700.00INR / Pkg of 500
Buy It Together
Current Item
Cole-Parmer Cleanroom Foam Swab, Open-Hole Head, Notched, Light Green PP Handle; 500/PK
Cole-Parmer Cleanroom Foam Swab, Open-Hole Head, Notched, Light Green PP Handle; 500/PK
3367605
₹19,700.00INR / Pkg of 500
+ =
Total Price 28100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Chemical resistance with a variety of solutions
  • Polypropylene handle ensures no additional contaminants
  • Suitable for ISO Class 5 cleanroom
  • Packed in large bag containing 5 bags of 100 swabs
More About this Item
These cleanroom swabs are ideal for precise cleaning, application of liquids or lubricants, and surface sampling validation. The swabs allow you to reduce the potential for lint and other residues in the cleanroom. The polypropylene handle helps to reduce any additional contaminants.
Specifications
  • MaterialFoam
  • ColorLight Green
  • ComplianceISO Class 5 cleanroom
  • Swab Total Length (InchShort)43/64
  • Swab Total Length (cm)1.7
  • Length (in)3 21/64
  • Length (cm)8.45
  • DescriptionCleanroom Foam Swab, Open-Hole Head, Notched, Light Green PP Handle; 500/PK
  • WarrantyReplacement only