Cole-Parmer Chart Paper for Flatbed Recorders; 200 mm wide, 4 rolls/pk

Representative image only.

Cole-Parmer Chart Paper for Flatbed Recorders; 200 mm wide, 4 rolls/pk

Cole-Parmer | Item: 8055082

₹23,500.00INR / Pkg of 4

₹23,500.00INR / Pkg of 4

Buy It Together

Current Item

Cole-Parmer Chart Paper for Flatbed Recorders; 200 mm wide, 4 rolls/pk
Cole-Parmer Chart Paper for Flatbed Recorders; 200 mm wide, 4 rolls/pk
8055082

₹23,500.00INR / Pkg of 4
+ =
Total Price 294500.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Replacement paper for your Cole-Parmer recorders

Specifications
  • Chart TypeStrip
  • Chart Width (in)7.875
  • DescriptionChart Paper for Flatbed Recorders; 200 mm wide, 4 rolls/pk
Reviews
Q&A