Cole-Parmer® Economical 100- and 200-mm Flatbed Recorders

Cole-Parmer® flatbed recorders combine accuracy with value
  • Set full scale ranges from 1 mV to 20 V
  • 12 selectable chart speeds are crystal controlled for optimum accuracy
  • Reliable long-life and simple operation
Measure reliably using the recorders’ zero control, 12 selectable input ranges and adjustable 5:1 attenuation. Compact housing design maximizes your bench space.
271,000.00 - ₹458,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chart Speed Chart Width (in) Number of Channels
Availability Pricing
COMPARE
1.0, 2.0, 5.0, 10, 20, 30 cm/min; 1.0, 2.0, 5.0, 10, 20, 30 cm/hr (user selectable) 4.75 1
₹271,000.00INR / Each
COMPARE
1.0, 2.0, 5.0, 10, 20, 30 cm/min; 1.0, 2.0, 5.0, 10, 20, 30 cm/hr (user selectable) 4.75 2
Call For Price
COMPARE
1.0, 2.0, 5.0, 10, 20, 30 cm/min; 1.0, 2.0, 5.0, 10, 20, 30 cm/hr (user selectable) 7.875 1
₹311,000.00INR / Each
COMPARE
1.0, 2.0, 5.0, 10, 20, 30 cm/min; 1.0, 2.0, 5.0, 10, 20, 30 cm/hr (user selectable) 7.875 2
₹458,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat