Cole-Parmer CELLROLL System, Culture Bottle/Tube Roller, 2.0 to 6.0 rpm; 90-260 VAC

Representative image only.

Cole-Parmer CELLROLL System, Culture Bottle/Tube Roller, 2.0 to 6.0 rpm; 90-260 VAC

Cole-Parmer | Item: 1305002

Easy to install, just click and roll

₹1,563,000.00INR / Each

₹1,563,000.00INR / Each

Buy It Together

Current Item

Cole-Parmer CELLROLL System, Culture Bottle/Tube Roller, 2.0 to 6.0 rpm; 90-260 VAC
Cole-Parmer CELLROLL System, Culture Bottle/Tube Roller, 2.0 to 6.0 rpm; 90-260 VAC
1305002

₹1,563,000.00INR / Each
+
Cole-Parmer FlexiRoll Digital Tube Roller Shaker; 120 VAC
0439744

₹379,000.00INR / Each
+ =
Total Price 2198000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for vaccine, insect, and antibody cell culture production
  • Advance remote functions and modular design allow easy cell culture scale-up
  • 18 month warranty

More About this Item

CELLROLL systems are programmable, modular roller systems that accommodate roller bottles with culture volumes from 300 to 400 mL. Systems operate by remote control and feature two ranges of rotating speeds. The control panel features a digital display, timer, and alarm; program for direction of rotation, bottle angle, velocity, and interval cycles. Systems are CO2 and moisture resistant.

Systems include two decks that hold four roller bottles and can be expanded to accommodate up to 16 bottles (total of eight decks). Fits vertically or horizontally inside a CO2 incubator. Control unit can be used outside the incubator for simple and flexible programming. One deck measures 11-5/8"L x14-3/16"W x 9-¼"H.

18 Month Warranty
Specifications
  • Number of Bottles Accepted4
  • Bottle Speed (rpm)2.0 to 6.0
  • Power (VAC)90 to 260
  • Capacity (mL)400
  • DescriptionCELLROLL system, 2.0 to 6.0 rpm, 90-260 VAC
  • Warranty18 months
Reviews
Q&A