Cole-Parmer BNC-to-Spade Lug Adapter
Representative image only.
Cole-Parmer BNC-to-Spade Lug Adapter
Cole-Parmer | Item: 0599490
Get the most out of water analysis equipment
₹6,100.00INR / Each
₹6,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer BNC-to-Spade Lug Adapter
Cole-Parmer BNC-to-Spade Lug Adapter
0599490
₹6,100.00INR / Each
+ +
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Transmitter
0580214
₹46,300.00INR / Each
+ + =
Total Price 216300.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Replace or upgrade electrodes, probes, cells, and other accessories
  • 1 year warranty
More About this Item
Water analysis accessories for Cole-Parmer® meters. Many of these items are compatible with other brands of meter.
1 Year Warranty
Specifications
  • Compatible WithTo connect any electrode with BNC to any controller with a spade lug strip
  • DescriptionBNC-to-Spade Lug Adapter