Cole-Parmer BioSafe BioSafe Serological Pipettes, 25 mL; 150/Cs

Representative image only.

Cole-Parmer BioSafe BioSafe Serological Pipettes, 25 mL; 150/Cs

Cole-Parmer | Item: 1300254

Filtered for sample integrity and user safety

₹8,000.00INR / Case of 150

₹8,000.00INR / Case of 150

Buy It Together

Current Item

Cole-Parmer BioSafe BioSafe Serological Pipettes, 25 mL; 150/Cs
Cole-Parmer BioSafe BioSafe Serological Pipettes, 25 mL; 150/Cs
1300254

₹8,000.00INR / Case of 150
+
Cole-Parmer Acrylic Pipette Organizing Rack; Blue
0439507

₹23,900.00INR / Each
=
Total Price 31900.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Eliminate cross-contamination — unique barrier filter prevents over-aspiration
  • Polystyrene pipettes are sterile, individually wrapped, and disposable
  • Ideal for use with potentially hazardous or dangerous biological fluids
  • Use in place of filtered pipette tips

More About this Item

These polystyrene serological pipettes feature a special filter plug to lock in fluid and vapors if over aspiration occurs. They also protect electronic pipettors from contamination and cross-contamination of samples from over-aspiration — promoting user safety and sample integrity.

Specifications
  • SterileYes
  • AutoclavableNo
  • Max Volume (mL)25
  • Graduation Interval0.2 mL
  • Accuracy (%)±0.5 mL
  • Tip StyleStandard
  • Body MaterialPolystyrene
  • CalibrationTo deliver (TD)
  • DescriptionBioSafe Serological Pipettes, 25 mL; 150/Cs
Reviews
Q&A