Cole-Parmer 8 Channel Mechanical Adjustable-Volume Pipette, 30 to 300 µL
Representative image only.
Cole-Parmer 8 Channel Mechanical Adjustable-Volume Pipette, 30 to 300 µL
Cole-Parmer | Item: 2450121
Ergonomic finger grip for comfortable use
₹30,800.00INR / Each
for an additional
₹110,000.00INR / Each
for an additional
₹119,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹30,800.00INR / Each
Buy It Together
Current Item
Cole-Parmer 8 Channel Mechanical Adjustable-Volume Pipette, 30 to 300 µL
Cole-Parmer 8 Channel Mechanical Adjustable-Volume Pipette, 30 to 300 µL
2450121
₹30,800.00INR / Each
+
Cole-Parmer EX Adjustable Volume Pipette, 0.1-2.0 uL; Each
0790901
₹66,600.00INR / Each
+
InnoCal NIST-Traceable Calibration; Pipette, 8 Channel
1710402
₹110,000.00INR / Each
+
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109
₹8,400.00INR / Each
+ +
Cole-Parmer Pipette Tip, 100 to 1000 µL, Bulk; 1000/BG
2450261
₹5,700.00INR / Bag of 1000
+ =
Total Price 574800.00 ADD 7 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Color-coding drop and removal tool, calibration/disassembling tool and silicon grease, along with O-rings, sample tips and calibration report.
Key Features
  • Features click-stop volume settings for simple and quick use
  • Ergonomic, lightweight push button design makes them easy to operate
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume displayed through handgrip window for quick glance convenience
  • Volume: 30 to 300 µL
  • 3 year warranty
More About this Item
These pipettes are offered in a wide-range of adjustable volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.
3 Year WarrantyConformite EuropeenneISO 9001:2008 Certified Supplier
Specifications
  • AutoclavableYes
  • Max Volume (µL)300
  • Min Volume (µL)30
  • Increments (µL)1
  • Accuracy (%)±0.8
  • Number Of Channels8
  • Tip EjectorYes
  • Color CodeBlue
  • Description8 Channel Mechanical Adjustable-Volume Pipette, 30 to 300 µL; 1/EA