Cole-Parmer 2650 Geno/Grinder® Homogenizer Cryo Adapter for 96-Well Microplate with Round Wells, Aluminum; 2/PK
Representative image only.
Cole-Parmer 2650 Geno/Grinder® Homogenizer Cryo Adapter for 96-Well Microplate with Round Wells, Aluminum; 2/PK
Cole-Parmer SamplePrep | Mfr: 2650| Item: 0457582
Get the most out of your Cole-Parmer® homogenizer
₹96,500.00INR / Pkg of 2
₹96,500.00INR / Pkg of 2
Buy It Together
Current Item
Cole-Parmer 2650 Geno/Grinder® Homogenizer Cryo Adapter for 96-Well Microplate with Round Wells, Aluminum; 2/PK
Cole-Parmer 2650 Geno/Grinder® Homogenizer Cryo Adapter for 96-Well Microplate with Round Wells, Aluminum; 2/PK
0457582
₹96,500.00INR / Pkg of 2
+ + + + + + =
Total Price 278100.00 ADD 7 ITEMS TO CART
Product Description
Key Features
  • Choose from a wide range of accessories for your homogenizer
  • Compatible with 04500-32 microplate
  • Insert in microplate wells to keep samples cold during tissue homogenization and cell lysis
More About this Item
Optimize your homogenizer with these quality accessories.
Use cryo adapter to increase yields while grinding samples at sub-zero temperatures. Used two at a time in the clamp assembly to keep it in balance.
Specifications
  • Use with GeneratorsDeep-well microplate 04500-32
  • MaterialAluminum
  • DescriptionGeno/Grinder® Homogenizer Cryo Adapter for 96-Well Microplate with Round Wells, Aluminum; 2/PK