Casella DBadge2Plus Noise Dosimeter Kit with 3 Meters

Representative image only.

Casella DBadge2Plus Noise Dosimeter Kit with 3 Meters

Casella | Mfr: DBADGE2PLUS/K3| Item: 5053663

The simplest way to obtain and record personal noise exposure

Call For Price

for an additional
₹38,500.00INR / Each
for an additional
₹50,200.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Casella DBadge2Plus Noise Dosimeter Kit with 3 Meters
Casella DBadge2Plus Noise Dosimeter Kit with 3 Meters
5053663

Call For Price
+
InnoCal NIST-Traceable Calibration; Sound Meter, Type II
1710153

₹38,500.00INR / Each
=
Total Price 38500.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Transmit sound level measurements wirelessly to increase productivity
  • Monitor multiple employees' noise exposure remotely and simultaneously
  • Select the appropriate hearing protection equipment—measurements are compliant with various regulation agencies
  • Securely mounts to the shoulder for worry-free, hands-free monitoring
  • Have confidence in your measurements—integral motion sensor expresses motion index in % of time
  • Kits come with everything needed to get started—add single dosimeters as needed
  • Includes three dosimeters, docking station/calibrator, charger, USB cable for direct data transfer to PC, Insight data analysis and summary software, a case to house up to 10 dosimeters, and access to the free Airwave app for your handheld device
  • 2 year warranty

More About this Item

Transmit sound level measurements through Bluetooth® 4.0 wireless connectivity to increase productivity—no need to stop working to download readings! Dose, status, level alerts and more can be accessed through the free Airwave™ app and sent directly to your Apple or Android enabled device. The dBadge2 noise dosimeters capture every possible noise exposure related parameter in a single measurement run, and make it available for subsequent download and analysis. Email summary data, photographs, and even notes for review or inclusion in reports.
Ensure the correct hearing protection is chosen for each application and location. The measurements taken are in accordance with CFR 1910.95, ISO 9612:2009, and UK Guidance document L108. The LCD displays current noise levels in color-coded format for easy interpretation of alarm conditions at a glance. Use the preset alarm limits to adhere to OSHA, ISO, MSHA, and ACGIH guidelines, or set your own.
Each personal noise dosimeter comes with two mounting clips, one at the top and one at the bottom, for secure shoulder attachment and features a reliable and robust 1/2" diameter microphone with windscreen. Order a kit to get started and add additional units as needed to cover an entire fleet!

dBadge2Plus Personal Noise Dosimeter Kits allow remote start, stop, or pause of a measurement run without interrupting the user—ideal for hazardous locations, positions, and filed data capture.
Additionally, these dosimeters have the option of recording audio events and notes. Store up to 15 seconds of voice recording at the start of a measurement run to assist with validation. Exposure limits may also be set to record the actual audio conditions if limits are exceeded. Audio recordings are saved in .WAV format and may be accessed through the included PC software.

Audio event recording: Settable level will trigger recording at 8 kHz sampling rate in a 16-bit compressed .WAV format. Playback is accessed through the PC software or other media player. Each audio event records for 15 seconds and includes a 3 second pre-trigger buffer to aid identification of the start of the noise event. Audio event trigger sources include LAF, LAS, LCPk, and LZPk.

Audio notes: Optional audio note of up to 15 seconds attached to the start of each measurement run.


Please visit http://www.casellasolutions.com/uk/en/support/dbadge2.aspx to download the latest software and utilities.

Two year warranty
Specifications
  • Decibel (Db) Low Range (dB)54
  • Decibel (Db) High Range (dB)140.3
  • Resolution (dB)0.1
  • WeightingA, C, Z (type 2)
  • Criterion Level70 to 90 dB (1 dB increments)
  • ComplianceANSI S1.25-1991 (R2007); IEC 61252 Ed. 1.1 (2002-3)
  • MemoryInternal USB Flash Memory (300 hours of run data, 1 second profile data, up to 90 minutes of event audio recording)
  • Display96 x 64 pixel full color OLED with adjustable contrast
  • Battery Type1 rechargeable NiMH (included)
  • Length (in)2 1/8
  • Height (in)3 3/8
  • Width (in)2 3/16
  • Length (cm)5.4
  • Height (cm)8.5
  • Width (cm)5.5
  • DescriptionNoise Dosimeter Kit with 3 Meters
Reviews
Q&A