InnoCal A2LA ISO17025 Accredited Calibration; Sound Meter, Type II
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration; Sound Meter, Type II
InnoCal | Item: 1750099
NIST-traceable sound calibrations--service available for sound-level meters (types I & II), noise dosimeters, sound filters, and sound-level calibrators
₹50,200.00INR / Each
₹50,200.00INR / Each
Buy It Together
Current Item
InnoCal A2LA ISO17025 Accredited Calibration; Sound Meter, Type II
InnoCal A2LA ISO17025 Accredited Calibration; Sound Meter, Type II
1750099
₹50,200.00INR / Each
+
Casella DBadge2 Noise Dosimeter; Meter Only
5053651
Call For Price
=
Total Price 50200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • A2LA Accredited Calibration. View our scope of accreditation
  • 1 year warranty
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005
Specifications
  • Calibration TypeA2LA ISO17025 Accredited Environmental Monitoring
  • Instrument Only CalibrationYes
  • Number Of Calibration Test Points2 Points per Range of the instrument
  • DescriptionA2LA ISO17025 Accredited Calibration; Sound Meter, Type II