Casella CEL-350IS Individual Intrinsically Safe micro noise badge, with mounting clips

Representative image only.

Casella CEL-350IS Individual Intrinsically Safe micro noise badge, with mounting clips

Casella | Mfr: dBadge2IS| Item: 9893335

Easily position on clothing or hard hats

₹515,000.00INR / Each

for an additional
₹38,500.00INR / Each
for an additional
₹50,200.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹515,000.00INR / Each

Buy It Together

Current Item

Casella CEL-350IS Individual Intrinsically Safe micro noise badge, with mounting clips
Casella CEL-350IS Individual Intrinsically Safe micro noise badge, with mounting clips
9893335

₹515,000.00INR / Each
+
InnoCal NIST-Traceable Calibration; Sound Meter, Type II
1710153

₹38,500.00INR / Each
=
Total Price 553500.00 ADD 2 ITEMS TO CART
Product Description

Includes

mounting clips and one internal rechargeable NiMH battery.

Key Features

  • Lightweight less than 2.5 oz
  • Small size—only 2.8" long, 1.7" wide and 2" tall
  • Quick charge in less than 1.5 hours from completely empty
  • Store all results for later review
  • Calculate all the popular noise dose indices
  • Time history profiling of 4 results at fixed 1 min intervals
  • Measure 3 dB and 5 dB exchange rates simultaneously
  • Lockable in operation mode
  • Direct automatic calibration with standard acoustic
  • calibrator (1⁄2" cavity)
  • Intrinsically safe model (98933-35) allows use in hazardous areas—these units have worldwide ATEX approval

More About this Item

The CEL350 has all the advantages of earlier badge designs, such as small size and light weight, and yet it still provides all of the benefits that users have come to expect from a personal noise dosimeter. The CEL350 is fully
featured and able to satisfy all current noise measurement protocols during a single run—an ideal instrument for both experienced and new users of dosimeters. Unit stores all results for later investigation and reporting. Easy, push-button setup.

Conformite Europeenne
Specifications
  • Decibel (Db) Low Range (dB)65
  • Decibel (Db) High Range (dB)140
  • Resolution (dB)0.1
  • Accuracy±1.5 dB
  • WeightingA (rms values); Z, C (peak measurements)
  • Criterion Level70 to 90 dB (1 dB increments)
  • Exchange Rate5 and 3 dB (simultaneously)
  • Threshold Level70 to 90 dB (1 dB increments)
  • FunctionLavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose%
  • ComplianceIEC 61252: 2002, BS EN 61252: 1997, ANSI S1.25 - 1992
  • OutputInfra-red to USB (PC) cable to Insight Software
  • Data LoggingYes
  • MemoryAt least 180 hours at 4 profiles every minute gives remaining memory recording time in hours when switched on
  • DisplayLCD
  • Battery Type1 rechargeable NiMH (included)
  • Height (in)2 51/64
  • Width (in)1 45/64
  • Depth (in)2
  • Height (cm)7.2
  • Width (cm)4.2
  • Depth (cm)5.2
  • Includesmouting clips
  • DescriptionMicro Noise Badge Intrinsically Safe with Mounting Clips and Windscreenr Only
Reviews
Q&A