Burkle 9622-0400 Funnel stand for 4 funnels, with four funnel holders for 40-180 mm
Representative image only.
Burkle 9622-0400 Funnel stand for 4 funnels, with four funnel holders for 40-180 mm
Burkle | Mfr: 9622-0400| Item: 0604441
Keep your hands free while using filters
₹79,600.00INR / Each
₹79,600.00INR / Each
Buy It Together
Current Item
Burkle 9622-0400 Funnel stand for 4 funnels, with four funnel holders for 40-180 mm
Burkle 9622-0400 Funnel stand for 4 funnels, with four funnel holders for 40-180 mm
0604441
₹79,600.00INR / Each
+ =
Total Price 91400.00 ADD 2 ITEMS TO CART
Product Description
Includes
06044-39 includes a 2-place base, 17-3/4" (45 cm) H stand rod, two funnel holders, and two standard funnel reducers. 06044-41 includes a 4-place base, 17-3/4" (45 cm) H stand rod, four funnel holders, and four standard funnel reducers.
Key Features
  • Chrome-plated steel and polypropylene construction resist chemicals
  • Compatible with 1.5- to 7-in (4- to 18-cm) diameter funnels
  • Adjust the height up to 17-3/4" (45 cm)
  • No clamps to tighten/untighten—innovative magnet design
  • Order additional funnel holders to change capacity and funnel type
  • 15 month warranty
More About this Item
Funnel stands come in either a 2- or 4-place design, and include corresponding funnel holders and standard adapters. Order Imhoff sedimentation funnel adapters separately.
Adjust the funnel support height with one hand—magnets have replaced screw-style clamps.
15 Month Warranty
Specifications
  • Clamp MaterialPolypropylene
  • Rod MaterialChrome-plated steel
  • Base MaterialPolypropylene
  • Clamp TypeFunnel
  • ColorWhite
  • DescriptionFunnel Stand, Four-Place with Clamps; 40-180 mm Diameter
  • Warranty15 month