Burkle 9622-0001 Funnel holder with reducing insert; for funnels 40-180 mm
Representative image only.
Burkle 9622-0001 Funnel holder with reducing insert; for funnels 40-180 mm
Burkle | Mfr: 9622-0001| Item: 0604438
Replacement clamps for your pipette or funnel stand
₹9,300.00INR / Each
₹9,300.00INR / Each
Product Description
Key Features
  • Magnets replace screw-type clamps for easy, one-handed adjustments
  • 15 month warranty
More About this Item
Clamps are compatible with 2- and 4-place funnel stands, as well as the pipette stand. Convert or increase your lab stand capacity.
15 Month Warranty
Specifications
  • Clamp MaterialPolypropylene
  • Clamp TypeFunnel
  • ColorWhite
  • DescriptionFunnel Clamp, Standard with Insert; 40-180 mm Diameter
  • Warranty15 month