Branson Temperature Probe for Digital Sonifiers, 200 and 400 W Models Only
Representative image only.
Branson Temperature Probe for Digital Sonifiers, 200 and 400 W Models Only
Branson | Mfr: 200-060-022R| Item: 0471567
Customize, upgrade, or replace parts
₹190,000.00INR / Each
₹190,000.00INR / Each
Buy It Together
Current Item
Branson Temperature Probe for Digital Sonifiers, 200 and 400 W models only
Branson Temperature Probe for Digital Sonifiers, 200 and 400 W models only
0471567
₹190,000.00INR / Each
+ + +
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
0471040
₹17,900.00INR / Each
=
Total Price 207900.00 ADD 4 ITEMS TO CART
Product Description
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Temperature probe allows for sample temperature monitoring and provides feedback necessary for the digital ultrasonic processor models to control sample temperature.
Specifications
  • DescriptionTemperature Probe for Digital Sonifiers, 200 and 400 W Models Only