Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
Representative image only.
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
Cole-Parmer | Item: 0471040
For threaded probes only
₹17,900.00INR / Each
₹17,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
0471040
₹17,900.00INR / Each
+ + =
Total Price 49700.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Autoclavable
  • 3 year warranty
More About this Item
The replaceable tip is for use with threaded probes and is fabricated from titanium alloy Ti-6Al-4V.
Use with threaded probe 04710-31.
3 Year Warranty
Specifications
  • Tip Diameter (in)0.5
  • Tip Diameter (cm)1.27
  • DescriptionThreaded Ultrasonic Probe Tip, Flat; 1/2" Dia