Branson Tapered Microtip for 1/2" Sonifier Tapped Horns, 1/4" dia
Representative image only.
Branson Tapered Microtip for 1/2" Sonifier Tapped Horns, 1/4" dia
Branson | Mfr: 101-148-070| Item: 0471585
Customize, upgrade, or replace parts
₹128,000.00INR / Each
₹128,000.00INR / Each
Buy It Together
Current Item
Branson Tapered Microtip for 1/2" Sonifier Tapped Horns, 1/4" dia
Branson Tapered Microtip for 1/2" Sonifier Tapped Horns, 1/4" dia
0471585
₹128,000.00INR / Each
+ + +
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
0471040
₹17,900.00INR / Each
=
Total Price 145900.00 ADD 4 ITEMS TO CART
Product Description
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Microtips for 1/2" diameter tapped horns feature titanium construction and a tapered shape. With higher amplitude and intensity, these tips are restricted to 70% (or lower) converter power and may not be operated in air, or a fracture may result.
Specifications
  • Tip Diameter (in)0.25
  • DescriptionTapered Microtip for 1/2" Sonifier Tapped Horns, 1/4" dia