Branson Microtip for Ultrasonic Processors, 3/32" dia
Representative image only.
Branson Microtip for Ultrasonic Processors, 3/32" dia
Branson | Mfr: 109-122-1066| Item: 0471540
Expand your ultrasonic processing capabilities
₹237,000.00INR / Each
₹237,000.00INR / Each
Buy It Together
Current Item
Branson Microtip for Ultrasonic Processors, 3/32" dia
Branson Microtip for Ultrasonic Processors, 3/32" dia
0471540
₹237,000.00INR / Each
+ + + =
Total Price 426000.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Get the most out of your Branson ultrasonic processor with these parts and accessories.
Microtips feature titanium construction and a tapered shape. With higher amplitude and intensity, these tips are restricted to 70% (or lower) converter power and may not be operated in air, or a fracture may result.
Specifications
  • Capacity (mL)Up to 1
  • Compatible WithBranson SFX150
  • Diameter (InchShort)3/32
  • DescriptionMicrotip for Ultrasonic Processors, 3/32" dia