Branson High-Gain Horn for Sonifiers, 3/4" dia, Stepped and Solid
Representative image only.
Branson High-Gain Horn for Sonifiers, 3/4" dia, Stepped and Solid
Branson | Mfr: 101-147-035R| Item: 0471558
Customize, upgrade, or replace parts
₹160,000.00INR / Each
₹160,000.00INR / Each
Buy It Together
Current Item
Branson High-Gain Horn for Sonifiers, 3/4" dia, stepped and solid
Branson High-Gain Horn for Sonifiers, 3/4" dia, stepped and solid
0471558
₹160,000.00INR / Each
+ + +
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
0471040
₹17,900.00INR / Each
=
Total Price 177900.00 ADD 4 ITEMS TO CART
Product Description
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Horns are fabricated from titanium. Available in different styles and sizes to suit a wide array of applications.
Specifications
  • Min Sample Size (mL)25
  • Max Sample Size (mL)500
  • Tip Diameter (in)0.75
  • DescriptionHigh-Gain Horn for Sonifiers, 3/4" dia, Stepped and Solid