Branson Flow Through Cup Horn for Sonifiers, 3" dia

Representative image only.

Branson Flow Through Cup Horn for Sonifiers, 3" dia

Branson | Mfr: 101-147-048| Item: 0471564

Customize, upgrade, or replace parts

₹812,000.00INR / Each

₹812,000.00INR / Each

Buy It Together

Current Item

Branson Flow Through Cup Horn for Sonifiers, 3" dia
Branson Flow Through Cup Horn for Sonifiers, 3" dia
0471564

₹812,000.00INR / Each
+ + +
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
0471040

₹17,900.00INR / Each
=
Total Price 829900.00 ADD 4 ITEMS TO CART
Product Description

More About this Item

Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.

Horns are fabricated from titanium. Available in different styles and sizes to suit a wide array of applications.

Specifications
  • Min Sample Size (mL)0.3
  • DescriptionFlow Through Cup Horn for Sonifiers, 3" dia
Reviews
Q&A