Branson Double Step Microtip f/ Direct Connection with Sonifier Converter, 1/8" dia
Representative image only.
Branson Double Step Microtip f/ Direct Connection with Sonifier Converter, 1/8" dia
Branson | Mfr: 101-063-212| Item: 0471586
Customize, upgrade, or replace parts
₹201,000.00INR / Each
₹201,000.00INR / Each
Buy It Together
Current Item
Branson Double Step Microtip f/ Direct Connection with Sonifier Converter, 1/8" dia
Branson Double Step Microtip f/ Direct Connection with Sonifier Converter, 1/8" dia
0471586
₹201,000.00INR / Each
+ + +
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
0471040
₹17,900.00INR / Each
=
Total Price 218900.00 ADD 4 ITEMS TO CART
Product Description
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Horns are fabricated from titanium. Available in different styles and sizes to suit a wide array of applications. The two-piece double-stepped microtip connects directly to the converter via a coupler, and fits into small-diameter tubes at a greater depth than standard microtips.
Specifications
  • Min Sample Size (mL)0.1
  • Max Sample Size (mL)10
  • Tip Diameter (in)0.125
  • DescriptionDouble Step Microtip f/ Direct Connection with Sonifier Converter, 1/8" dia