Branson Disruptor Horn for Sonifiers, 3/8" dia, Stepped and Solid
Representative image only.
Branson Disruptor Horn for Sonifiers, 3/8" dia, Stepped and Solid
Branson | Mfr: 101-147-039| Item: 0471553
Customize, upgrade, or replace parts
₹205,000.00INR / Each
₹205,000.00INR / Each
Buy It Together
Current Item
Branson Disruptor Horn for Sonifiers, 3/8" dia, stepped and solid
Branson Disruptor Horn for Sonifiers, 3/8" dia, stepped and solid
0471553
₹205,000.00INR / Each
+ + +
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
0471040
₹17,900.00INR / Each
=
Total Price 222900.00 ADD 4 ITEMS TO CART
Product Description
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Horns are fabricated from titanium. Available in different styles and sizes to suit a wide array of applications.
Specifications
  • Min Sample Size (mL)5
  • Max Sample Size (mL)100
  • Tip Diameter (in)0.375
  • DescriptionDisruptor Horn for Sonifiers, 3/8" dia, Stepped and Solid