Branson Disruptor Horn for Sonifiers, 3/4" dia, Stepped and Tapped
Representative image only.
Branson Disruptor Horn for Sonifiers, 3/4" dia, Stepped and Tapped
Branson | Mfr: 611-005-021| Item: 0471559
Customize, upgrade, or replace parts
₹231,000.00INR / Each
₹231,000.00INR / Each
Buy It Together
Current Item
Branson Disruptor Horn for Sonifiers, 3/4" dia, stepped and tapped
Branson Disruptor Horn for Sonifiers, 3/4" dia, stepped and tapped
0471559
₹231,000.00INR / Each
+
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
0471040
₹17,900.00INR / Each
+ + =
Total Price 248900.00 ADD 4 ITEMS TO CART
Product Description
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Horns are fabricated from titanium. Available in different styles and sizes to suit a wide array of applications, choose a tapped tip if you need to use a threaded microtip for low volumes.
Specifications
  • Min Sample Size (mL)25
  • Max Sample Size (mL)500
  • Tip Diameter (in)0.75
  • DescriptionDisruptor Horn for Sonifiers, 3/4" dia, Stepped and Tapped