Branson Disruptor Horn for Sonifiers, 1/2" dia, Stepped and Tapped
Representative image only.
Branson Disruptor Horn for Sonifiers, 1/2" dia, Stepped and Tapped
Branson | Mfr: 101-147-037R| Item: 0471551
Customize, upgrade, or replace parts
₹212,000.00INR / Each
₹212,000.00INR / Each
Buy It Together
Current Item
Branson Disruptor Horn for Sonifiers, 1/2" dia, stepped and tapped
Branson Disruptor Horn for Sonifiers, 1/2" dia, stepped and tapped
0471551
₹212,000.00INR / Each
+ + +
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
0471040
₹17,900.00INR / Each
=
Total Price 229900.00 ADD 4 ITEMS TO CART
Product Description
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Horns are fabricated from titanium. Available in different styles and sizes to suit a wide array of applications, choose a tapped tip if you need to use a threaded microtip for low volumes.
Specifications
  • Min Sample Size (mL)10
  • Max Sample Size (mL)250
  • Tip Diameter (in)0.5
  • DescriptionDisruptor Horn for Sonifiers, 1/2" dia, Stepped and Tapped