Branson Continuous Flow Attachment for Sonifiers, Stainless Steel
Representative image only.
Branson Continuous Flow Attachment for Sonifiers, Stainless Steel
Branson | Mfr: 100-146-171| Item: 0471568
Expand your ultrasonic processing capabilities
₹676,000.00INR / Each
₹676,000.00INR / Each
Buy It Together
Current Item
Branson Continuous Flow Attachment for Sonifiers, stainless steel
Branson Continuous Flow Attachment for Sonifiers, stainless steel
0471568
₹676,000.00INR / Each
+ + + =
Total Price 865000.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Get the most out of your Branson ultrasonic processor with these parts and accessories.
Cells create controlled environments for samples. The continuous flow attachment works with low-viscosity fluids only, and is ideal for large batches and for recirculating a sample to improve homogenization. This attachment is jacketed in order to keep the sample cool, especially important for sample recirculating.
Specifications
  • MaterialStainless Steel
  • Compatible WithBranson Sonifiers
  • DescriptionContinuous Flow Attachment for Sonifiers, Stainless Steel