BrandTech 704261 Remote Dispensing System for Dispensette® S/S Organic, Standard Supply
Representative image only.
BrandTech 704261 Remote Dispensing System for Dispensette® S/S Organic, Standard Supply
BrandTech | Mfr: 704261| Item: 0791508
Accurately dispense from drums and bulk refills
₹190,000.00INR / Each
₹190,000.00INR / Each
Buy It Together
Current Item
BrandTech 704261 Remote Dispensing System for Dispensette® S/S Organic, Standard Supply
BrandTech 704261 Remote Dispensing System for Dispensette® S/S Organic, Standard Supply
0791508
₹190,000.00INR / Each
+ =
Total Price 341000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
PTFE plug-in adapter†, PP GL 45/32 thread adapter, PP GL 32/28 thread adapter, PP stain-free grip, ETFE locking screws (2), PP wall-mount unit, 7.6 mm OD x 3 m L FEP filling tube, 6.9 mm OD x 47 cm L FEP filling tube, PTFE mounting screw, ETFE coupling with ball valve, PTFE drum adapter with 3/4" inner thread, nonsterile, and 3 µm nonsterile membrane filter
Key Features
  • Filter minimizes risk of contamination
  • Mount on a wall, ring stand, or lab furniture
More About this Item
Dispensing accurate volumes from drums and bulk containers has never been easier. Designed to work with Dispensette® S and S Organic dispensers, the remote dispensing system can be mounted up to 30 feet (10 meters) away from the source. A quick-release connector with integrated valves allow quick changing of the container. A built-in filter minimizes contamination risk of high-purity reagents. Easily mount on a wall, ring stand, or lab furniture—maximum delivery height is 4 ft (1.2 m).
Dispensette® dispenser not included.
†Plug-in adapter is required to use 10 mL and smaller Dispensette® dispensers.
Specifications
  • Compatible WithDispensette S and Dispensette S Organic dispensers
  • DescriptionRemote Dispensing System for Dispensette® S/S Organic