BrandTech® Dispensette® S Bottletop Dispensers
Best choice for most general purpose dispensing
  • Proven floating piston design provides one handed operation even for large volumes
  • Numerous safety features
  • Chemically resistant materials, such as PTFE, ETFE, PFA, FEP, Al203, platinum-iridium and borosilicate glass
  • Hinged screw cap hinges away from dispensing when removed
  • Available with priming valve
This fourth-generation Dispensette® S bottletop dispenser is an excellent choice for general dispensing. A reinforced discharge tube with swing-away screw cap improves safety while an improved fluid path requires even lower operating forces. A safety discharge valve reduces the chance of dispensing unless the discharge tube is mounted properly.
Choose a dispenser with a recirculation valve if you want chemicals to route back into the reagent bottle during the priming step. Analog models feature adjustable volume selection with interior scalloped track to provide reproducibe settings.
Dispensette® S Dispensers support a wide range of applications. Dispense bases, buffers, many acids and organic solvents. Not for use with hydrofluoric acid.
151,000.00 - ₹343,000.00INR / Each
33 variations of this product are available.
See All Product Options Below
33 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Dispense Volume (mL) Reproducibility
Availability Pricing
BrandTech Dispensette® S Bottletop Dispenser, Fixed Volume, with Standard Valve, 1 mL 0791511 Mfr # 4600210
COMPARE
±0.6% 1 ≤ 0.2%
₹151,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Fixed Volume, with Recirculation Valve, 1 mL 0791512 Mfr # 4600211
COMPARE
±0.6% 1 ≤ 0.2%
₹158,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Fixed Volume, with Standard Valve, 2 mL 0791513 Mfr # 4600220
COMPARE
±0.5% 2 ≤ 0.1%
₹151,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Fixed Volume, with Recirculation Valve, 2 mL 0791514 Mfr # 4600221
COMPARE
±0.5% 2 ≤ 0.1%
₹158,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Fixed Volume, with Standard Valve, 5 mL 0791515 Mfr # 4600230
COMPARE
±0.5% 5 ≤ 0.1%
₹151,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Fixed Volume, with Recirculation Valve, 5 mL 0791516 Mfr # 4600231
COMPARE
±0.5% 5 ≤ 0.1%
₹158,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Fixed Volume, with Standard Valve, 10 mL 0791517 Mfr # 4600240
COMPARE
±0.5% 10 ≤ 0.1%
₹151,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Fixed Volume, with Recirculation Valve, 10 mL 0791518 Mfr # 4600241
COMPARE
±0.5% 10 ≤ 0.1%
₹158,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Standard Valve, 0.1-1 mL 0791521 Mfr # 4600100
COMPARE
±0.6% 1 ≤ 0.2%
₹151,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Standard Valve, 0.2-2 mL 0791523 Mfr # 4600120
COMPARE
±0.5% 2 ≤ 0.1%
₹151,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Recirculation Valve, 0.2-2 mL 0791524 Mfr # 4600121
COMPARE
±0.5% 2 ≤ 0.1%
₹158,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Standard Valve, 0.5-5 mL 0791525 Mfr # 4600130
COMPARE
±0.5% 5 ≤ 0.1%
₹151,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Recirculation Valve, 0.5-5 mL 0791526 Mfr # 4600131
COMPARE
±0.5% 5 ≤ 0.1%
₹158,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Standard Valve, 1-10 mL 0791527 Mfr # 4600140
COMPARE
±0.5% 10 ≤ 0.1%
₹151,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with recirculation valve, 1-10 mL 0791528 Mfr # 4600141
COMPARE
±0.5% 10 ≤ 0.1%
₹158,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Standard Valve, 2.5-25 mL 0791529 Mfr # 4600150
COMPARE
±0.5% 25 ≤ 0.1%
₹209,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Reciculating Valve, 2.5-25 mL 0791530 Mfr # 4600151
COMPARE
±0.5% 25 ≤ 0.1%
₹216,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Standard Valve, 5-50 mL 0791531 Mfr # 4600160
COMPARE
±0.5% 50 ≤ 0.1%
₹215,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with recirculation valve, 5-50 mL 0791532 Mfr # 4600161
COMPARE
±0.5% 50 ≤ 0.1%
₹222,000.00INR / Each
BrandTech Dispensette® S Bottletop Dispenser, Analog Adjustable, with Standard Valve, 10-100 mL 0791533 Mfr # 4600170
COMPARE
±0.5% 100 ≤ 0.1%
₹334,000.00INR / Each
Showing 1 - 20 of 33