Advantec Manifold, Stainless Steel, 6 stations
Representative image only.
Advantec Manifold, Stainless Steel, 6 stations
Advantec | Mfr: G1300| Item: 3521157
Create your own vacuum filtration systems
₹1,245,000.00INR / Each
₹1,245,000.00INR / Each
Buy It Together
Current Item
Advantec Manifold, Stainless Steel, 6 stations
Advantec Manifold, Stainless Steel, 6 stations
3521157
₹1,245,000.00INR / Each
+ =
Total Price 1399000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose PVC or stainless steel construction
  • Control each branch independently with 3-way PTFE valves
  • 3 month warranty
3 Month Warranty
Specifications
  • Body MaterialStainless Steel
  • DescriptionManifold, Stainless Steel, 6 stations