Advantec GA100142MM Grade GA100 Glass Fiber Filters; 142mm dia; 100/box

Representative image only.

Advantec GA100142MM Grade GA100 Glass Fiber Filters; 142mm dia; 100/box

Advantec | Mfr: GA100142MM| Item: 0665757

For analytical or gravimetric studies

₹33,000.00INR / Pkg of 100

₹33,000.00INR / Pkg of 100

Buy It Together

Current Item

Advantec GA100142MM Grade GA100 Glass Fiber Filters; 142mm dia; 100/box
Advantec GA100142MM Grade GA100 Glass Fiber Filters; 142mm dia; 100/box
0665757

₹33,000.00INR / Pkg of 100
+ =
Total Price 37900.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

General purpose; filtering cells and precipitated proteins; air pollution monitoring.

1 Year Warranty
Specifications
  • MaterialGlass Microfiber
  • Diameter (mm)142
  • Diameter (cm)14.2
  • DescriptionGrade GA100 Glass Fiber Filters; 142mm dia; 100/box
  • Warranty1 year
Reviews
Q&A