3M 11326-00000-20 Safety Eyewear, Clear frame, Clear lens
Representative image only.
3M 11326-00000-20 Safety Eyewear, Clear frame, Clear lens
3M | Mfr: 11326-00000-20| Item: 8647270
Lightweight design, feels almost like its not there
₹800.00INR / Each
₹800.00INR / Each
Buy It Together
Current Item
3M 11326-00000-20 Safety Eyewear, Clear frame, Clear lens
3M 11326-00000-20 Safety Eyewear, Clear frame, Clear lens
8647270
₹800.00INR / Each
+ +
Cole-Parmer Safety Wash Bottle, LDPE, Acetone, 500mL (16oz); 6/PK
0625640
₹14,700.00INR / Box of 6
+ =
Total Price 43500.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Sleek, 9.5-base lens provides superior protection while offering unobstructed view
More About this Item
Tough polycarbonate construction is designed to resist impacts and block 99.9% of UV rays. Features contoured, flexible temples, and universal-fit nosebridge.
The hard coat on the lens guards against scratches, static, and chemical attack.
Compliance: ANSI Z87.1-2003
Specifications
  • Frame ColorClear
  • Lens ColorClear
  • Lens CoatingUltra-dura® (hard coating)
  • ComplianceANSI Z87.1-2003
  • DescriptionSafety Eyewear, Clear frame, Clear lens