Cole-Parmer® Class A, Type 1 Glass Erlenmeyer Flasks
Class A, Type 1 laboratory Erlenmeyer flasks meet ASTM E1404, DIN 12380
  • Durable heavy-duty beaded rim design
  • Withstand temperatures from -50 to 550 °C (-58 to 1022 °F)
  • Easy-to-read graduations
  • Autoclavable
Ideal for mixing solutions, titration experiments, yeast culturing, chemistry experiments and more, these Erlenmeyer flasks are manufactured from Class A, Type 1 borosilicate glass that meets ASTM E1404, DIN 12380 specifications. They have a heavy-duty beaded rim to help prevent breakage and approximate graduations indicated in easy-to-read, durable white enamel. Able to withstand temperatures of -50 to 550°C (-58 to 1022°F), they are autoclavable for sterilization.
5,600.00 - ₹10,700.00INR / Pkg of 6
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Min Graduation (mL)
Availability Pricing
COMPARE
100 20 20
₹8,200.00INR / Pkg of 12
COMPARE
250 50 50
₹8,500.00INR / Pkg of 8
COMPARE
500 100 100
₹8,300.00INR / Pkg of 8
COMPARE
1000 100 100
₹10,700.00INR / Pkg of 6
COMPARE
2000 250 250
₹5,600.00INR / Each
COMPARE
3000 500 500
₹8,400.00INR / Each