Cell-Based Assays The Steps of Assay Development and Screening in Early Drug Discovery
Assay development and screening are crucial steps in the process of drug discovery, where scientists aim to identify and develop new drugs for the treatment of various diseases.
Secondary substance screening and lead optimization
Measure the activity of a substance, such as a drug candidate, in a biological system
Create and optimize the tests to reliably and accurately measure the desired biological activity
Assay Development and Screening
Integral parts of drug discovery that involve creating reliable tests and rapidly evaluating large numbers of compounds to identify potential drug candidates for further development. These processes help researchers identify potential drug candidates and understand their effects on specific biological targets.
An assay is a test or experiment designed to measure the activity of a substance, such as a drug candidate, in a biological system. Assay development involves creating and optimizing these tests to reliably and accurately measure the desired biological activity.
Types of Assays
- Biochemical Assays: Measure the interaction between a drug candidate and a specific biomolecule (e.g., enzyme, receptor).
- Biophysical Assays: Focus on studying the physical properties of biological molecules, such as proteins or nucleic acids, to understand their behavior and interactions in response to potential drug compounds.
- Cell-Based Assays: Assess the effects of a drug candidate on living cells, providing a more physiologically relevant environment.