Jenway 74-Series Spectrophotometers Spectrum Scan Video
How to Perform a Spectrum Scan on Cloud-enabled Spectrophotometers
Description
A short how-to video demonstrating how to peform a spectrum scan using the new Jenway 74 Series Visible and UV/Visible scanning spectrophotometers
The Steps
  1. To Start touch Spectrum
  2. Touch start wavelength
  3. Use the keypad to enter the required wavelength
  4. Touch the scan interval
  5. Select the required ineteral
  6. Following a baseline scan touch sample to perform the spectrum scan
Once the spectrum scan is complete, the peaks and valleys can be identified and analyzed.
The 74 Series spectrophotometers are covered with a 3 year warranty.
Jenway Scanning UV/Visible Spectrophotometer with CPLive Cloud Data Storage
Browse the 74 Series Spectrophotometers from Jenway
See all Jenway spectrophometers products