Dataloggers help 30-year-old school overcome mold problems
Dataloggers help 30-year-old school
overcome mold problems
With outdoor humidity rates near a constant 100% during the summer months, Houston is quickly becoming known as one of the world's moldiest cities. Many of the area's insurance carriers have seen a significant increase in mold claims in recent years, and in the past two years alone, carriers have paid more than $1 billion in mold settlements.
While virtually all types of buildings and structures in Houston are susceptible to mold growth, area schools are among the most affected. Those allowing fresh air in on a regular basis are particularly affected.
Such was the case with Montgomery Elementary School in North Houston. With tropical rain forest-like air constantly infiltrating the classrooms of this 30-year-old structure, signs of mold growth started to surface in late 2001.
'Like a dirty wet sock'
"There were some visible indications of mold in certain areas, and some of the classrooms smelled like a dirty wet sock," explained Kyle Jeane, a HVAC technician with Houston-based Hal Watson A/C Co. "In addition, kids with asthma were becoming affected."
In addition to exacerbating asthma, the mycotoxins that mold spores produce are known to be responsible for other problems from suppression of the immune system to neurological damage. School officials asked Jeane to investigate the situation. More specifically, he was asked to gather data on classroom humidity levels and, based on this information, make recommendations on a dehumidification solution. To do so, he would need to set up dehumidifiers in a few classrooms, and provide analytical data showing the difference in humidity between a classroom with a dehumidifier, and one without.
"While you could easily feel the difference in humidity with dehumidification, the school's budget approval process made it necessary to show actual results based on analysis," said Jeane. "This data would ultimately have to be presented to school architects, engineers, and the school board itself."
To record and analyze the classroom climate conditions, Jeane relied on HOBO(r) H8 dataloggers from Onset Computer Corporation. These compact, battery-powered devices are used for continuous monitoring of temperature, rh, light intensity, and other environmental conditions. He was able to hang a logger on the wall in both the humid and dehumidified classrooms using a paperclip on a nail. Once mounted, the loggers started recording temperature, rh, and dewpoint at 30-minute intervals.
After logging classroom conditions for three days, Jeane removed the loggers and offloaded the data onto a PC. For graphing and analysis, he used BoxCar Pro, a Windows-based software program that accompanies the dataloggers.
"The software was very user-friendly and it only took a few minutes to get all the data collected into nice graphical charts," he explained. "From the charts, we could immediately see a dramatic split in terms of humidity rates in the two classrooms. The classroom with the dehumidifier was running at 45% humidity, while the other was closer to 95%. This was tangible evidence that our dehumidifiers were able to override the high dewpoints and pull the humidity down to a safe level."
Mold growth now a nonissue
Jeane proceeded to print out the graphs and presented his findings to school officials. Upon reviewing the results, the school quickly approved funding for 15 Thermastor 150H humidifiers, which were installed soon thereafter. The school classrooms now have dial-stats on the wall, encased in a lock-box, and each is set to 45% humidity. Mold growth has become a nonissue, and the school air feels and smells clean and fresh.
More Details or Order Online:
Temperature/Humidity Dataloggers with Alarm Output